missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM189 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 220.00 - € 518.00
Specifications
| Antigen | TMEM189 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645051
|
Novus Biologicals
NBP2-94610-0.02ml |
0.02 mL |
€ 220.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611401
|
Novus Biologicals
NBP2-94610-0.1ml |
0.1 mL |
€ 518.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM189 Polyclonal antibody specifically detects TMEM189 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TMEM189 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KUA, transmembrane protein 189, UBE2V1, ubiquitin-conjugating enzyme E2 variant 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 171-270 of human TMEM189 (NP_954580.1). FVFCLIIFGTFTNQIHKWSHTYFGLPRWVTLLQDWHVILPRKHHRIHHVSPHETYFCITTGWLNYPLEKIGFWRRLEDLIQGLTGEKPRADDMKWAQKIK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 387521 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title