Learn More
TOP2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-58728
 View more versions of this product
Product Code. 18615816
  • ONLINE EXCLUSIVE PRICE
    € 792.00
    € 748.65 / 100µL
    Save € 43.35
    5% Off
Description
TOP2B Polyclonal antibody specifically detects TOP2B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TOP2B
Polyclonal
Western Blot 0.04-0.4 μg/mL
antigen MLAA-44, DNA topoisomerase 2-beta, DNA topoisomerase II beta, DNA topoisomerase II, 180 kD, DNA topoisomerase II, beta isozyme, EC 5.99.1.3, top2beta, TOPIIB, topo II beta, topoisomerase (DNA) II beta (180kD), topoisomerase (DNA) II beta 180kDa, topoisomerase II beta, topoisomerase IIb, U937 associated antigen
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence: DKVPSKTVAAKKGKPSSDTVPKPKRAPKQKKVVEAVNSDSDSEFGIPKKTTTPKGKGRGAKKRKASGSENEGDYNPGRKTSKTTSKKPKKTSFDQDSDVDIFPSDFPTEPPSLPRTGRARKEVKYFAESDEEEDDVDF
100 μg
DNA Repair, DNA replication Transcription Translation and Splicing
7155
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Affinity purified