missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 748.65
Specifications
| Antigen | TOP2B |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637168
|
Novus Biologicals
NBP2-58728-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615816
|
Novus Biologicals
NBP2-58728 |
100 μg |
€ 792.00 € 748.65 / 100µL Save € 43.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TOP2B Polyclonal antibody specifically detects TOP2B in Human samples. It is validated for Western BlotSpecifications
| TOP2B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.2) and 40% Glycerol | |
| 7155 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen MLAA-44, DNA topoisomerase 2-beta, DNA topoisomerase II beta, DNA topoisomerase II, 180 kD, DNA topoisomerase II, beta isozyme, EC 5.99.1.3, top2beta, TOPIIB, topo II beta, topoisomerase (DNA) II beta (180kD), topoisomerase (DNA) II beta 180kDa, topoisomerase II beta, topoisomerase IIb, U937 associated antigen | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence: DKVPSKTVAAKKGKPSSDTVPKPKRAPKQKKVVEAVNSDSDSEFGIPKKTTTPKGKGRGAKKRKASGSENEGDYNPGRKTSKTTSKKPKKTSFDQDSDVDIFPSDFPTEPPSLPRTGRARKEVKYFAESDEEEDDVDF | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title