missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOP3A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5121539

 View more versions of this product

Product Code. 17772934

  • € 184.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DNA topoisomerases are nuclear enzymes that regulate the topological structure of DNA by transiently breaking and rejoining DNA strands. Although DNA topoisomerase I (Topo I) and Topo II relax both positive and negative supercoils, Topo III relaxes only negative supercoils. Topo IIIalpha exists as a long and a short isoform, which are produced by alternative splicing. Topo IIIalpha, which localizes to the nucleolus, is constitutively expressed and remains at high levels throughout the cell cycle in HL-60 cells. Topo IIIbeta exists as three isoforms,beta-1,beta -2 and beta-3, also produced by alternative splicing. Topo IIIbeta-1 is expressed in testis, heart, and skeletal muscle, whereas Topo IIIbeta-2 is expressed in thymus, kidney and pancreas.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TOP3A
Polyclonal
Unconjugated
Top3a
DNA topoisomerase 3-alpha; DNA topoisomerase III alpha; Top IIIa; TOP3; TOP3A; topo III-alpha; topoisomerase (DNA) 3 alpha; topoisomerase (DNA) III alpha; topoisomerase 3 alpha; ZGRF7; zinc finger, GRF-type containing 7
Rabbit
Antigen affinity chromatography, Protein A
RUO
7156
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q13472
Top3a
Lys35-Ser179 (Accession Q13472), with N-terminal His Tag.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.