Learn More
TORC3/CRTC3 Antibody (10G3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26644-50ul
 View more versions of this product
Product Code. 30043818
  • € 343.00 / 50µL
Description
TORC3/CRTC3 Monoclonal antibody specifically detects TORC3/CRTC3 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TORC3/CRTC3
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
64784
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
ELISA, Immunohistochemistry
10G3
ELISA, Immunohistochemistry 1:50-1:200
CREB regulated transcription coactivator 3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3
A synthesized peptide derived from Human TORC3/CRTC3 [UniProt Q6UUV7]
50 μL