missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TORC3/CRTC3 Antibody (10G3), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | TORC3/CRTC3 |
|---|---|
| Clone | 10G3 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043818
|
Novus Biologicals
NBP3-26644-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043386
|
Novus Biologicals
NBP3-26644-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TORC3/CRTC3 Monoclonal antibody specifically detects TORC3/CRTC3 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| TORC3/CRTC3 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 64784 | |
| IgG | |
| Affinity purified |
| 10G3 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CREB regulated transcription coactivator 3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3 | |
| A synthesized peptide derived from Human TORC3/CRTC3 [UniProt Q6UUV7] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title