Learn More
Ansell™ TouchNTuff™ 83-300 Gloves
Shop All Ansell Products
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Unit Size:
200 pairs
Description
- Compatible with Class 100 (ISO 5) / Grade A Cleanroom Environments
- Added comfort with second skin feel
- Latex-free polyisoprene performance
- Prevents Type I allergies
- Extended cuff
- Cuff style is beaded with Surefit™ Technology
- Freeedom from holes 0.65 AQL
- Double glove recommended
- Internal glove surface is coated
- Anatomic with curved fingers
Specifications
Specifications
| Certifications/Compliance | ASTM D2577, CE Marked CE0493 |
| Chlorinated | Yes |
| Description | Smooth external glove surface |
| Applications | Laboratory |
| Color | White |
| Latex Free | Yes |
| Material | Polyisoprene |
| Product Type | Disposable Gloves |
| Size | 7.5 |
| Thickness (English) | 0.25 mil |
| Show More |