missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53I13 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94004-0.1ml
This item is not returnable.
View return policy
Description
TP53I13 Polyclonal antibody specifically detects TP53I13 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TP53I13 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DSCP1Damage-stimulated cytoplasmic protein 1, tumor protein p53 inducible protein 13, tumor protein p53-inducible protein 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 180-310 of human TP53I13 (NP_612358.3). RSWRPPGTEVTSQGPRQPSSSGAKRRRLRAALGPQPTRSALRFPSASPGSLKAKQSMAGIPGRESNAPSVPTVSLLPGAPGGNASSRTEAQVPNGQGSPGGCVCSSQASPAPRAAAPPRAARGPTPRTEEA | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 90313 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction