Learn More
Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1023X
This item is not returnable.
View return policy
Description
TRA-1-60 Monoclonal Antibody for Flow, ICC/IF, Western Blot
TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
Specifications
| TRA-1-60 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| Mouse | |
| Affinity chromatography - MBP | |
| RUO | |
| Human | |
| Antibody | |
| IgM |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| TRA-1-60 | |
| Unconjugated | |
| TRA160; tra60 | |
| Human embryonal carcinoma cell line 2102Ep. | |
| 20 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |