missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60)

Mouse Monoclonal Antibody
€ 227.00 - € 447.00
Specifications
| Antigen | TRA-1-60 |
|---|---|
| Clone | TRA-1-60 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12817752
![]() |
Invitrogen™
MA1023 |
100 μg |
€ 447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
12897742
|
Invitrogen™
MA1023X |
20 μL |
€ 227.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRA-1-60 Monoclonal Antibody for Flow, ICC/IF, Western Blot
TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.Specifications
| TRA-1-60 | |
| 1 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| Human embryonal carcinoma cell line 2102Ep. | |
| Antibody |
| TRA-1-60 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgM | |
| Human | |
| TRA160; tra60 | |
| Primary | |
| Affinity chromatography - MBP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title