missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-2 Antibody (33A1293) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-56715SS
This item is not returnable.
View return policy
Description
TRAF-2 Monoclonal specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRAF-2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 2 ug/ml, Immunohistochemistry 2:20-2:1000, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Galen et al (2008)) | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HeLa, an approx. 57 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 33A1293 | |
| Unconjugated | |
| Q12933 | |
| TRAF2 | |
| Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2. | |
| 0.025 mg | |
| Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
| 7186 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction