missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-2 Antibody (33A1293) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
€ 200.00 - € 486.00
Specifications
| Antigen | TRAF-2 |
|---|---|
| Clone | 33A1293 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 2:20-2:1000, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Galen et al (2008)) |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075345
|
Novus Biologicals
NB100-56715SS |
0.025 mg |
€ 200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18708883
|
Novus Biologicals
NB100-56715 |
0.1 mg |
€ 486.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF-2 Monoclonal specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TRAF-2 | |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
| Q12933 | |
| 7186 | |
| Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 33A1293 | |
| Western Blot 2 ug/ml, Immunohistochemistry 2:20-2:1000, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Galen et al (2008)) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| TRAF2 | |
| IgG1 κ | |
| Protein G purified | |
| In HeLa, an approx. 57 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title