missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-86913
This item is not returnable.
View return policy
Description
TRAF-2 Polyclonal specifically detects TRAF-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRAF-2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TRAF2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KLEAKYLCSACRNVLRRPFQAQCGHRYCSFCLASILSSGPQNCAACVHEGIYEEGISILESSSAFPDNAARREVESLPAVCPSDGCTWKGTLKEYESCHEGRCPLMLTECPACKGLVRLGEKERHLEHECPERSLSCRHCRAPC | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
| 7186 | |
| Human, Mouse | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering