missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 302.00 - € 456.00
Specifications
| Antigen | TRAF-2 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18454491
|
Novus Biologicals
NBP1-86913-25ul |
25 μL |
€ 302.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18226558
|
Novus Biologicals
NBP1-86913 |
0.1 mL |
€ 456.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF-2 Polyclonal specifically detects TRAF-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TRAF-2 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| TRAF2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7186 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KLEAKYLCSACRNVLRRPFQAQCGHRYCSFCLASILSSGPQNCAACVHEGIYEEGISILESSSAFPDNAARREVESLPAVCPSDGCTWKGTLKEYESCHEGRCPLMLTECPACKGLVRLGEKERHLEHECPERSLSCRHCRAPC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title