missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAM2 Recombinant Rabbit Monoclonal Antibody (24GB1960)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557289
This item is not returnable.
View return policy
Description
TRAM2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Necessary for collagen type I synthesis. May couple the activity of the ER Ca(2+) pump SERCA2B with the activity of the translocon. This coupling may increase the local Ca(2+) concentration at the site of collagen synthesis, and a high Ca(2+) concentration may be necessary for the function of molecular chaperones involved in collagen folding. Required for proper insertion of the first transmembrane helix N-terminus of TM4SF20 into the ER lumen, may act as a ceramide sensor for regulated alternative translocation (RAT).
Specifications
| TRAM2 | |
| Recombinant Monoclonal | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15035, Q924Z5 | |
| TRAM2 | |
| A synthesized peptide derived from human TRAM2 (aa 250-300). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1960 | |
| Unconjugated | |
| TRAM2 | |
| C330003D03Rik; KIAA0057; LOC100361830; tram2; TRAM-like protein; Translocating chain-associated membrane protein 2; translocating chain-associating membrane protein 2; translocation associated membrane protein 2; wu:fb20c11; wu:fi23a12; zgc:111902; zgc:55869; zgc:76937 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100361830, 170829, 9697 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction