missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAM2 Recombinant Rabbit Monoclonal Antibody (24GB1960)

Rabbit Recombinant Monoclonal Antibody
€ 146.00 - € 500.00
Specifications
| Antigen | TRAM2 |
|---|---|
| Clone | 24GB1960 |
| Concentration | 0.15 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275088
![]() |
Invitrogen™
MA557289 |
100 μL |
€ 500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275358
|
Invitrogen™
MA557290 |
20 μL |
€ 146.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAM2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Necessary for collagen type I synthesis. May couple the activity of the ER Ca(2+) pump SERCA2B with the activity of the translocon. This coupling may increase the local Ca(2+) concentration at the site of collagen synthesis, and a high Ca(2+) concentration may be necessary for the function of molecular chaperones involved in collagen folding. Required for proper insertion of the first transmembrane helix N-terminus of TM4SF20 into the ER lumen, may act as a ceramide sensor for regulated alternative translocation (RAT).Specifications
| TRAM2 | |
| 0.15 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15035, Q924Z5 | |
| 100361830, 170829, 9697 | |
| A synthesized peptide derived from human TRAM2 (aa 250-300). | |
| Antibody |
| 24GB1960 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| TRAM2 | |
| C330003D03Rik; KIAA0057; LOC100361830; tram2; TRAM-like protein; Translocating chain-associated membrane protein 2; translocating chain-associating membrane protein 2; translocation associated membrane protein 2; wu:fb20c11; wu:fi23a12; zgc:111902; zgc:55869; zgc:76937 | |
| TRAM2 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title