Learn More
Invitrogen™ TRAM2 Recombinant Rabbit Monoclonal Antibody (24GB1960)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557290
 View more versions of this product
Product Code. 30275358
  • € 146.00 / 20µL
Description
TRAM2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Necessary for collagen type I synthesis. May couple the activity of the ER Ca(2+) pump SERCA2B with the activity of the translocon. This coupling may increase the local Ca(2+) concentration at the site of collagen synthesis, and a high Ca(2+) concentration may be necessary for the function of molecular chaperones involved in collagen folding. Required for proper insertion of the first transmembrane helix N-terminus of TM4SF20 into the ER lumen, may act as a ceramide sensor for regulated alternative translocation (RAT).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRAM2
Recombinant Monoclonal
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q15035, Q924Z5
TRAM2
A synthesized peptide derived from human TRAM2 (aa 250-300).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1960
Unconjugated
TRAM2
C330003D03Rik; KIAA0057; LOC100361830; tram2; TRAM-like protein; Translocating chain-associated membrane protein 2; translocating chain-associating membrane protein 2; translocation associated membrane protein 2; wu:fb20c11; wu:fi23a12; zgc:111902; zgc:55869; zgc:76937
Rabbit
Affinity chromatography
RUO
100361830, 170829, 9697
-20°C