Learn More
trans,trans-2,4-Dodecadienal, 90+%, remainder mainly trans, cis isomer
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.863 |
| Boiling Point | 130°C (6 mmHg) |
| Flash Point | 104°C (219°F) |
| Refractive Index | 1.51 |
| Quantity | 1 g |
| Beilstein | 1708096 |
| Formula Weight | 180.29 |
| Percent Purity | ≥90% |
| Assay | remainder mainly trans, cis isomer |
| Chemical Name or Material | trans,trans-2,4-Dodecadienal |