missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transducin alpha Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3504-0.025mg
This item is not returnable.
View return policy
Description
Transducin alpha Polyclonal specifically detects Transducin alpha in Mouse, Porcine, Bovine, Sheep samples. It is validated for Western Blot.
Specifications
| Transducin alpha | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GNAT1 | |
| Synthetic Peptide: K(18) L K E D A E K D A R T V K(31) C | |
| 0.025 mg | |
| Signal Transduction, Vision | |
| 2779 | |
| Mouse | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL | |
| CSNBAD3, GBT1, GNATR, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 1, guanine nucleotide-binding protein G(t) subunit alpha-1, guanine nucleotide-binding protein G(T), alpha-1 subunit, rod-type transducin alpha subunit, Transducin alpha-1 chain, transducin, rod-specific | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects Rod Transducin alpha from bovine outer rod segments and sheep retinal extract. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction