missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transducin alpha Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00
Specifications
| Antigen | Transducin alpha |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Transducin alpha Polyclonal specifically detects Transducin alpha in Mouse, Porcine, Bovine, Sheep samples. It is validated for Western Blot.Specifications
| Transducin alpha | |
| Western Blot 1 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction, Vision | |
| PBS with 0.05% Sodium Azide | |
| 2779 | |
| Synthetic Peptide: K(18) L K E D A E K D A R T V K(31) C | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse | |
| CSNBAD3, GBT1, GNATR, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 1, guanine nucleotide-binding protein G(t) subunit alpha-1, guanine nucleotide-binding protein G(T), alpha-1 subunit, rod-type transducin alpha subunit, Transducin alpha-1 chain, transducin, rod-specific | |
| GNAT1 | |
| IgG | |
| Affinity Purified | |
| Detects Rod Transducin alpha from bovine outer rod segments and sheep retinal extract. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title