Learn More
Novus Biologicals™ Transglutaminase 2/TGM2 Antibody (zampilimab) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28563-1mg
 View more versions of this product
Product Code. 30046030
  • € 920.00 / 1mg
Description
Transglutaminase 2/TGM2 Monoclonal antibody specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transglutaminase 2/TGM2
Monoclonal
Unconjugated
P21980
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG4
Flow Cytometry, ELISA, Functional Assay
zampilimab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2
TGM2/ Transglutaminase 2
1 mg
Apoptosis, Immune System Diseases, Immunology, Signal Transduction
7052