missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM10 Monoclonal Antibody (K92040_6C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555634
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: LEESPTCPLC KEPFRPGSFR PNWQLANVVE NIERLQLVST LGLGEEDVCQ EHGEKIYFFC EDDEMQLCVV CREAGEHATH TMRFLEDAAA PYREQIHKCL KCLRKEREEI QEIQSRENKR
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
Specifications
| TRIM10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9UDY6 | |
| TRIM10 | |
| Recombinant human TRIM10 (immunogen range is 51-170). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K92040_6C6 | |
| Unconjugated | |
| TRIM10 | |
| AI324236; B30-RING finger protein; BB139825; Hematopoietic RING finger 1; Herf1; RFB30; RING finger protein 9; Rnf9; TRIM10; tripartite motif containing 10; tripartite motif protein 10; tripartite motif-containing 10; tripartite motif-containing protein 10; Zn-finger protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 10107, 294210 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction