missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM10 Monoclonal Antibody (K92040_6C6), Invitrogen™
Mouse Monoclonal Antibody
€ 438.00 - € 658.00
Specifications
| Antigen | TRIM10 |
|---|---|
| Clone | K92040_6C6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30206598
|
Invitrogen™
MA556014 |
200 μL |
€ 658.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30212594
|
Invitrogen™
MA555634 |
50 μL |
€ 438.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: LEESPTCPLC KEPFRPGSFR PNWQLANVVE NIERLQLVST LGLGEEDVCQ EHGEKIYFFC EDDEMQLCVV CREAGEHATH TMRFLEDAAA PYREQIHKCL KCLRKEREEI QEIQSRENKR
TRIM10 Monoclonal antibody specifically detects TRIM10 in Human, Rat samples. It is validated for Western BlotSpecifications
| TRIM10 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI324236; B30-RING finger protein; BB139825; Hematopoietic RING finger 1; Herf1; RFB30; RING finger protein 9; Rnf9; TRIM10; tripartite motif containing 10; tripartite motif protein 10; tripartite motif-containing 10; tripartite motif-containing protein 10; Zn-finger protein | |
| Recombinant human TRIM10 (immunogen range is 51-170). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TRIM10 |
| K92040_6C6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| Q9UDY6 | |
| 10107, 294210 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title