missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM24 Mouse anti-Human, Mouse, Clone: 1D6D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66324-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc binding domains (a RING, a B box type 1 and a B box type 2) and a coiled coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| TRIM24 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15164, Q64127 | |
| TRIM24 | |
| TRIM24 Fusion Protein Ag5417 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1D6D9 | |
| Unconjugated | |
| TRIM24 | |
| hTIF1, PTC6, RING finger protein 82, RNF82, TF1A, TIF1, TIF1 alpha, TIF1A, TIF1ALPHA, TRIM24, tripartite motif containing 24 | |
| Mouse | |
| Protein G | |
| RUO | |
| 21848, 8805 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction