missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM24 Mouse anti-Human, Mouse, Clone: 1D6D9, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | TRIM24 |
|---|---|
| Clone | 1D6D9 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858095
|
Proteintech
66324-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848095
|
Proteintech
66324-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc binding domains (a RING, a B box type 1 and a B box type 2) and a coiled coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| TRIM24 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hTIF1, PTC6, RING finger protein 82, RNF82, TF1A, TIF1, TIF1 alpha, TIF1A, TIF1ALPHA, TRIM24, tripartite motif containing 24 | |
| TRIM24 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1D6D9 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| O15164, Q64127 | |
| 21848, 8805 | |
| TRIM24 Fusion Protein Ag5417 | |
| Primary | |
| -20°C | |
| TRIM24 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title