missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM65 Monoclonal Antibody (K52045_14A2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556133
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: DAERLKREAQ LRASLEVTQQ QATQAEGQLL ELRKQSSQIQ NSACILASWV SGKFSSLLQA LEIQHTTALR SIEVAKTQAL AQARDEEQRL RVHLEAVARH GCRIRELLEQ VDEQTFLQES QLLQPPGPLG PLTPLQWDED QQLGDLKQLL SRLCGLLLEE GSHPGAPAKP VDLAPVEAPG
TRIM65 is a protein coding gene and gene ontology (GO) includes cytosol, nucleoplasm, nucleus, positive regulation of autophagy.
Specifications
| TRIM65 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q6PJ69 | |
| TRIM65 | |
| Recombinant human TRIM65 (immunogen range is 131-310). | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K52045_14A2 | |
| Unconjugated | |
| TRIM65 | |
| 4732463G12Rik; TRIM65; tripartite motif containing 65; tripartite motif-containing protein 65 | |
| Mouse | |
| Protein A | |
| RUO | |
| 201292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction