missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM65 Monoclonal Antibody (K52045_14A2), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | TRIM65 |
|---|---|
| Clone | K52045_14A2 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30210799
|
Invitrogen™
MA555753 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30199541
|
Invitrogen™
MA556133 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: DAERLKREAQ LRASLEVTQQ QATQAEGQLL ELRKQSSQIQ NSACILASWV SGKFSSLLQA LEIQHTTALR SIEVAKTQAL AQARDEEQRL RVHLEAVARH GCRIRELLEQ VDEQTFLQES QLLQPPGPLG PLTPLQWDED QQLGDLKQLL SRLCGLLLEE GSHPGAPAKP VDLAPVEAPG
TRIM65 Monoclonal antibody specifically detects TRIM65 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| TRIM65 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 4732463G12Rik; TRIM65; tripartite motif containing 65; tripartite motif-containing protein 65 | |
| Recombinant human TRIM65 (immunogen range is 131-310). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TRIM65 |
| K52045_14A2 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6PJ69 | |
| 201292 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title