Learn More
TRIM72 Mouse, anti-Human, CoraLite Plus 488, Clone: 5F2C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67403-100UL
Description
The TRIM72 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM72. This antibody recognizes human, pig, mouse antigen. The TRIM72 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| TRIM72 | |
| 5F2C11 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| hMG53, Mg53, Mitsugumin 53, TRIM72, tripartite motif containing 72 | |
| 100 μL | |
| 493829 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |