Learn More
TRIM72 Mouse, anti-Human, CoraLite Plus 488, Clone: 5F2C11, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | TRIM72 |
|---|---|
| Clone | 5F2C11 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087415
|
Proteintech
CL488-67403-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18015615
|
Proteintech
CL488-67403-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TRIM72 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM72. This antibody recognizes human, pig, mouse antigen. The TRIM72 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| TRIM72 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 493829 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 5F2C11 | |
| Immunofluorescence | |
| Liquid | |
| Human, Pig, Mouse | |
| hMG53, Mg53, Mitsugumin 53, TRIM72, tripartite motif containing 72 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.