missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkC Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A7F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66380-1-IG-20UL
This item is not returnable.
View return policy
Description
TRKC, a member of the insulin receptor subfamily of tyrosine protein kinases, is a receptor for neurotrophin-3 (NT-3). Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.Specifications
| TrkC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24786, Q03351, Q16288, Q6VNS1 | |
| Ntrk3 | |
| TrkC Fusion Protein Ag2635 | |
| 20 μL | |
| Primary | |
| Pig, Mouse, Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2A7F11 | |
| Unconjugated | |
| NTRK3 | |
| GP145 TrkC, gp145(trkC), NT 3 growth factor receptor, NTRK3, Trk C, TRKC, TrkC tyrosine kinase | |
| Mouse | |
| Protein G | |
| RUO | |
| 18213, 29613, 397511, 4916 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction