missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkC Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A7F11, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | TrkC |
|---|---|
| Clone | 2A7F11 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858205
|
Proteintech
66380-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848205
|
Proteintech
66380-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRKC, a member of the insulin receptor subfamily of tyrosine protein kinases, is a receptor for neurotrophin-3 (NT-3). Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.Specifications
| TrkC | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GP145 TrkC, gp145(trkC), NT 3 growth factor receptor, NTRK3, Trk C, TRKC, TrkC tyrosine kinase | |
| Ntrk3 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2A7F11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Pig, Mouse, Human, Rat | |
| P24786, Q03351, Q16288, Q6VNS1 | |
| 18213, 29613, 397511, 4916 | |
| TrkC Fusion Protein Ag2635 | |
| Primary | |
| -20°C | |
| NTRK3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title