Learn More
TRNT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand:  Novus Biologicals NBP1-86589
 View more versions of this product
Product Code. 18788503
  • € 557.00 / 0.10mL
Description
TRNT1 Polyclonal specifically detects TRNT1 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunofluorescence
Western Blot 0.04 to 0.4 μg/mL, Simple Western 1:8, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated
50 kDa
IgG
Polyclonal
TRNT1
Human, Rat