missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRNT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
€ 206.00 - € 557.00
Specifications
| Antigen | TRNT1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18443131
|
Novus Biologicals
NBP1-86589-25ul |
25 μL |
€ 206.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788503
|
Novus Biologicals
NBP1-86589 |
€ 557.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TRNT1 Polyclonal specifically detects TRNT1 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| TRNT1 | |
| Polyclonal | |
| Rabbit | |
| Human, Rat | |
| Q96Q11 | |
| 51095 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LDLRLKIAKEEKNLGLFIVKNRKDLIKATDSSDPLKPYQDFIIDSREPDATTRVCELLKYQGEHCLLKEMQQWSIPPFPVSGHDIR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 50 kDa |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CCA1, CCA-adding, CGI-47, EC 2.7.7.72, mitochondrial CCA-adding tRNA-nucleotidyltransferase, mt CCA-adding enzyme, mt tRNA adenylyltransferase, mt tRNA CCA-diphosphorylase, mt tRNA CCA-pyrophosphorylase, MtCCA, tRNA nucleotidyl transferase, CCA-adding, 1, tRNA-nucleotidyltransferase 1, mitochondrial | |
| TRNT1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human TRNT1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel