missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM8 Antibody (SR1110), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21832-50ul
This item is not returnable.
View return policy
Description
TRPM8 Monoclonal antibody specifically detects TRPM8 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| TRPM8 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1110 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
| A synthesized peptide derived from human TRPM8 (Uniprot #: Q7Z2W7) | |
| 50 μg | |
| Cancer, Tumor Suppressors | |
| 79054 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction