missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM8 Antibody (SR1110), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | TRPM8 |
|---|---|
| Clone | SR1110 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623444
|
Novus Biologicals
NBP3-21832-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671554
|
Novus Biologicals
NBP3-21832-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPM8 Monoclonal antibody specifically detects TRPM8 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| TRPM8 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
| A synthesized peptide derived from human TRPM8 (Uniprot #: Q7Z2W7) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1110 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 79054 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title