missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSCOT Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93061-0.02ml
This item is not returnable.
View return policy
Description
TSCOT Polyclonal antibody specifically detects TSCOT in Human, Mouse samples. It is validated for Western Blot
Specifications
| TSCOT | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| Solute carrier family 46 member 2, solute carrier family 46, member 2, thymic stromal co-transporter, thymic stromal cotransporter homolog, TSCOTLy110 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human SLC46A2 (NP_149040.3). MSPEVTCPRRGHLPRFHPRTWVEPVVASSQVAASLYDAGLLLVVKASYGTGGSSNHSASPSPRGALEDQQQRAISNFYIIYNLVVGLSPLLSAYGLGWLSDRYHRKISIC | |
| 0.02 mL | |
| Cancer | |
| 57864 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction