missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSCOT Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | TSCOT |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690151
|
Novus Biologicals
NBP2-93061-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673550
|
Novus Biologicals
NBP2-93061-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSCOT Polyclonal antibody specifically detects TSCOT in Human, Mouse samples. It is validated for Western BlotSpecifications
| TSCOT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 57864 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Solute carrier family 46 member 2, solute carrier family 46, member 2, thymic stromal co-transporter, thymic stromal cotransporter homolog, TSCOTLy110 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human SLC46A2 (NP_149040.3). MSPEVTCPRRGHLPRFHPRTWVEPVVASSQVAASLYDAGLLLVVKASYGTGGSSNHSASPSPRGALEDQQQRAISNFYIIYNLVVGLSPLLSAYGLGWLSDRYHRKISIC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title