missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSG101 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98994-100ul
This item is not returnable.
View return policy
Description
TSG101 Polyclonal antibody specifically detects TSG101 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| TSG101 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1500-1:50000, Immunoprecipitation 0.2-1 uL/mg of lysate | |
| ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TSG101. | |
| 100 μg | |
| Autophagy, Cancer, Microautophagy | |
| 7251 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction