missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSG101 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 271.00 - € 326.00
Specifications
| Antigen | TSG101 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1500-1:50000, Immunoprecipitation 0.2-1 uL/mg of lysate |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648105
|
Novus Biologicals
NBP2-98994-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628305
|
Novus Biologicals
NBP2-98994-50ul |
50 μg |
€ 271.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSG101 Polyclonal antibody specifically detects TSG101 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| TSG101 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Microautophagy | |
| 0.2 μm filtered solution in PBS | |
| 7251 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1500-1:50000, Immunoprecipitation 0.2-1 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TSG101. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts