missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC11 Antibody (27B4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26579-100ul
This item is not returnable.
View return policy
Description
TTC11 Monoclonal antibody specifically detects TTC11 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| TTC11 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 27B4 | |
| Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
| CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
| A synthesized peptide derived from Human TTC11 [UniProt Q9Y3D6] | |
| 100 μL | |
| Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
| 51024 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction