Learn More
TTL Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E5F8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66076-1-IG-20UL
 View more versions of this product
Product Code. 16837625
  • € 145.00 / 20µL
Description
Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.
Specifications
Specifications
TTL
Monoclonal
0.76 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P38160, P38585, Q8NG68, Q9QXJ0
Ttl
TTL Fusion Protein Ag4708
20 μL
Primary
Human, Rat, Pig, Mouse
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin)
2E5F8
Unconjugated
TTL
TTL, Tubulin tyrosine ligase, tubulin tyrosine ligase
Mouse
Protein A
RUO
150465, 171572, 445530, 69737
-20°C