missing translation for 'onlineSavingsMsg'
Learn More

TTL Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E5F8, Proteintech

Mouse Monoclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen TTL
Clone 2E5F8
Concentration 0.76 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16837625
View Documents
Proteintech
66076-1-IG-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16827625
View Documents
Proteintech
66076-1-IG-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.
Specifications

Specifications

TTL
0.76 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
TTL, Tubulin tyrosine ligase, tubulin tyrosine ligase
Ttl
IgG2b
Protein A
Antibody
2E5F8
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Rat, Pig, Mouse
P38160, P38585, Q8NG68, Q9QXJ0
150465, 171572, 445530, 69737
TTL Fusion Protein Ag4708
Primary
-20°C
TTL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.