Learn More
Fisherbrand™ Tube Rack for Shaking Incubators
Product Code. 16373236 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
1.5 to 2 mL Tubes
15 mL Tubes
15 mL, 50 mL Tubes
50 mL Tubes
500 mL Tubes
Tubes
Tilt Angle:
Horizontal
Vertical
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Tilt Angle unitSize
16373236 15 mL Tubes Vertical Each
16393226 15 mL, 50 mL Tubes Vertical Each
16363236 Tubes Vertical Each
16303246 50 mL Tubes Vertical Each
16323246 500 mL Tubes Vertical Each
16393236 1.5 to 2 mL Tubes Horizontal Each
16383236 15 mL Tubes Horizontal Each
16313246 50 mL Tubes Horizontal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
Product Code. 16373236
Only null left
Add to basket
Add to basket
Fisherbrand™ Tube Rack is designed to be used with a shaking incubator and can hold a variety of tube sizes.
  • For use with shaking incubator
  • Constructed of plastic, polypropylene, aluminum, or wire
  • Designed to hold tubes
Specifications
Tilt Angle Vertical
Holds 15 mL Tubes
Tube Capacity 15 mL
Product Type Tube Rack
Material Polypropylene
For Use With (Equipment) 16313226
No. of Positions 32
Packaging Quantity 1 Ea.