Learn More
Fisherbrand™ Tube-Shaped Stirrer Bars
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
9 mm
Unit Size:
Pack of 10
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| Dimensions (L x W x H) | 9 x 9 x 6mm |
| For Use With (Equipment) | Cuvettes |
| Height (Metric) | 6mm |
| Diameter (Metric) | 9 mm |
| Length (Metric) | 9 mm |
| Quantity | 5/Pk. |