missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Tuff Cut Nit Gloves
Click to view available options
Size:
7
8
9
10
Unit Size:
10 pairs
Description
- Knitted glove in Kevlar™, high tenacity polyamide, steel fiber.
- Knitted construction for hand conformity and dexterity.
- Polyurethane-coated palm for abrasion resistance.
- Nitrile foam coating for oil and grease resistance and improved flexibility over standard nitrile.
- Outstanding cutting resistance (Level 5).
- Suited for handling heavy parts with high cut risks in oily and/or greasy environments
- Light heat protection (up to 100°C).
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical Hazards, EN407-2004 Thermal risks, EC Category PPE 2, EC Certificate Number 0075/797/162/04/13/0287, EC Attestation AET_2232533_Honeywell 2013, EC Attestation Number 0075/797/162/04/13/0287 |
| Classification | Dexterity: 5 |
| Coated Area | Palm, Fingers |
| Description | Gauge 10 |
| Coating Material | Nitrile Foam |
| Color | Black |
| Cut Resistance Rating | 5 |
| Material | Aramid Fiber, Polyamide, Stainless Steel Fiber, Nitrile, Dupont™ Kevlar™ Aramid Fibers |
| Product Type | Gloves |
| Size | 9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction