missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Tuff Cut Nit Gloves

Product Code. p-7115900
Click to view available options
Size:
7
8
9
10
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 15706008

Brand: Honeywell Safety Products™ 223253308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Comfortable and flexible glove constructed of a variety of materials for superior resistance to cutting, puncture, oil and grease. Honeywell Safety Products™ Tuff Cut Nit Gloves are knitted with Kevlar™, high tenacity polyamide, and steel fiber, and coated in flexible nitrile foam.

  • Knitted glove in Kevlar™, high tenacity polyamide, steel fiber.
  • Knitted construction for hand conformity and dexterity.
  • Polyurethane-coated palm for abrasion resistance.
  • Nitrile foam coating for oil and grease resistance and improved flexibility over standard nitrile.
  • Outstanding cutting resistance (Level 5).
  • Suited for handling heavy parts with high cut risks in oily and/or greasy environments
  • Light heat protection (up to 100°C).
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical Hazards, EN407-2004 Thermal risks, EC Category PPE 2, EC Certificate Number 0075/797/162/04/13/0287, EC Attestation AET_2232533_Honeywell 2013, EC Attestation Number 0075/797/162/04/13/0287
Classification Dexterity: 5
Coated Area Palm, Fingers
Description Gauge 10
Coating Material Nitrile Foam
Color Black, Yellow
Cut Resistance Rating 5
Material Aramid Fiber, Polyamide, Stainless Steel Fiber, Nitrile, Dupont™ Kevlar™ Aramid Fibers
Product Type Gloves
Size 8
For Use With (Application) Automotive, Aviation, Building, Construction, General, Metal, Mining, Quarrying, Offshore, Paper and Ship Building industries.
Puncture Rating 3
Quantity 20/PK
Tear Rating 4
Length (Metric) 30 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.