Learn More
Invitrogen™ TXNDC5 Monoclonal Antibody (K52034_11G7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556138
Description
Sequence of this protein is as follows: AAAAAADGPP AADGEDGQDP HSKHLYTADM FTHGIQSAAH FVMFFAPWCG HCQRLQPTWN DLGDKYNSME DAKVYVAKVD CTAHSDVCSA QGVRGYPTLK LFKPGQEAVK YQGPRDFQTL ENWMLQTLNE EPVTPEPEVE PPSAPELKQG LYELSASNFE LHVAQGDHFI KFFAPWCGHC KALAPTWEQL ALGLEHSETV KIGKVDCTQH YELCSGNQVR GYPTLLWFRD GKKVDQYKGK RDLESLREYV
This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| TXNDC5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8NBS9 | |
| Txndc5 | |
| Recombinant human TXNDC5 (immunogen range is 41-290). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K52034_11G7 | |
| Unconjugated | |
| Txndc5 | |
| AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700 | |
| Mouse | |
| Protein A | |
| RUO | |
| 81567 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.