Learn More
Invitrogen™ TXNDC5 Monoclonal Antibody (K52034_11G7), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | TXNDC5 |
|---|---|
| Clone | K52034_11G7 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30200115
|
Invitrogen™
MA555758 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30210526
|
Invitrogen™
MA556138 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: AAAAAADGPP AADGEDGQDP HSKHLYTADM FTHGIQSAAH FVMFFAPWCG HCQRLQPTWN DLGDKYNSME DAKVYVAKVD CTAHSDVCSA QGVRGYPTLK LFKPGQEAVK YQGPRDFQTL ENWMLQTLNE EPVTPEPEVE PPSAPELKQG LYELSASNFE LHVAQGDHFI KFFAPWCGHC KALAPTWEQL ALGLEHSETV KIGKVDCTQH YELCSGNQVR GYPTLLWFRD GKKVDQYKGK RDLESLREYV
TXNDC5 Monoclonal antibody specifically detects TXNDC5 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| TXNDC5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700 | |
| Recombinant human TXNDC5 (immunogen range is 41-290). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Txndc5 |
| K52034_11G7 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8NBS9 | |
| 81567 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.