missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ Peristaltic Tubing E-LFL

Fits tightly to glass and metal, bends to small radius curves, is nontoxic and transparent. Tygon™ Peristaltic Tubing E-LFL has long life and won’t age or oxidize.

Brand:  Saint Gobain Life Sciences 9208909

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10778921

  • € 128.00 / Pack of 12
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

  • Comprised of Tygon™ flexible polymer
  • Translucent
  • Resistant to acids and alkalies
  • For use with specialized fluid transfer
Specifications

Specifications

Peristaltic Pump Tubing
1.02 mm
1.2 ft.
Peristaltic Pumps
Flexible
0.04 in.
Tygon E-LFL
38 cm
Acids, Alkalies
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers