missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TYKi Antibody (CL14479), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26857-25ul
This item is not returnable.
View return policy
Description
TYKi Monoclonal antibody specifically detects TYKi in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TYKi | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, cytidylate kinase 2, EC 2.7.4.14, mitochondrial, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member, TMPK2, TYKi, UMP-CMPK2 | |
| This antibody was developed against a Synthetic Peptide of Q5EBM0. Actual immunogen sequence is proprietary. | |
| 25 μL | |
| Protein Kinase | |
| 129607 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL14479 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction